Learn More
Medchemexpress LLC 3-(3-Trifluoromethylphenyl)propanol | 78573-45-2 | C10H11F3O | 5 G
SDP
Supplier:  Medchemexpress LLC HY760395G
Encompass_Preferred
3-(3-Trifluoromethylphenyl)propanol is a drug intermediate for the synthesis of various active compounds.
  • Drug intermediate for synthesis
  • For research use only
  • Store at 4°C, sealed, away from moisture
  • In solvent: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)
Catalog No. 50-004-34662