Learn More
Medchemexpress LLC 3-[(3R,4R)-1-[(3S)-3-cyclohexyl-3-hydroxypropyl]-3,4-dimethylpiperidin-4-yl]phenol | 119193-09-8 | 98.0% | 345.52 g/mol | C22H35NO2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1189495MG
Encompass_Preferred
LY255582 is a pan-opioid receptor antagonist research compound used to study opioid receptor signaling, feeding behavior, and addiction. Supplied as a laboratory reagent for research use only (not for human or veterinary use).
  • Pan-opioid antagonist active at mu, delta, and kappa receptors.
  • Chemical formula C22H35NO2; molecular weight 345.52 g/mol.
  • Supplied as a 5 mg solid with 98.0% purity (HPLC reported).
  • Used in pharmacology studies of appetite, addiction, and receptor binding.
  • Store desiccated at low temperature for stability.
Catalog No. 50-002-27399