Learn More
Medchemexpress LLC 3-[[4-(2-pyridinyl)-2-thiazolyl]amino]phenol | 315702-75-1 | MFCD01121976 | 100.0% | C14H11N3OS | 10MG
SDP
Supplier:  Medchemexpress LLC HY13103110MG
Encompass_Preferred
KCC-07 is a brain-penetrant small molecule inhibitor of MBD2 (methyl-CpG-binding domain protein 2) that prevents MBD2 binding to methylated DNA and activates BAI1-p53-p21 signaling to produce antiproliferative effects. It is supplied as a solid and as a 10 mM solution in DMSO, with reported high purity and recommended storage conditions for research use.
  • Inhibits MBD2 binding to methylated DNA.
  • Blood-brain barrier penetrant.
  • Activates BAI1-p53-p21 antiproliferative signaling.
  • High purity suitable for research applications.
  • Soluble at high concentration in DMSO.
  • Available as solid and as 10 mM DMSO solution.
  • Storage recommendations provided for long-term stability.
Catalog No. 50-002-61845