Learn More
Medchemexpress LLC 3-[4-(4-hydroxyphenyl)piperazin-1-yl]-1-phenylpyrrolidine-2,5-dione | 522649-59-8 | 99.7% | 351.4 g/mol | C20H21N3O3 | 100MG
SDP
Supplier:  Medchemexpress LLC HY124861100MG
Encompass_Preferred
Malic enzyme inhibitor ME1 is a small-molecule research reagent that potently inhibits cytosolic malic enzyme 1 (ME1) with an IC50 of 0.15 μM. The compound (CAS 522649-59-8) has formula C20H21N3O3, molecular weight 351.4 g/mol, and is supplied as a high-purity powder (≈99.7%) or as a 10 mM solution in DMSO for biochemical and cell-based studies.
  • Potent inhibition of ME1 (IC50 0.15 μM).
  • High purity suitable for research applications.
  • Available as powder and as a 10 mM solution in DMSO.
  • Multiple size options including 100 mg for scale-up studies.
  • Stable when stored under recommended frozen conditions.
  • Applicable to biochemical inhibition assays and cell-based metabolic studies.
Catalog No. 50-002-35719