Learn More
Medchemexpress LLC 3,4,5-trimethoxy-N-(2-(3-(piperazin-1-ylmethyl)imidazo[2,1-b][1,3]thiazol-6-yl)phenyl)benzamide | 925432-73-1 | 98.2% | 507.60 g·mol⁻1 | C26H29N5O4S | 50MG
SDP
Supplier:  Medchemexpress LLC HY12403750MG
Encompass_Preferred
SRT 1460 is a small-molecule activator of Sirtuin 1 (SIRT1) used in biochemical and cell-based research. It exhibits potent activation of SIRT1 with an EC1.5 of 2.9 μM and shows strong selectivity over SIRT2 and SIRT3. The compound is supplied as an off-white to light yellow solid with high purity for laboratory studies.
  • Potent SIRT1 activation (EC1.5 = 2.9 μM).
  • Selective versus SIRT2 and SIRT3 (EC1.5 > 300 μM).
  • High purity suitable for research (98.2%).
  • Molecular formula C26H29N5O4S; molecular weight 507.60 g·mol⁻1.
  • Off-white to light yellow solid physical form.
  • Stable under recommended storage for long-term use.
Catalog No. 50-002-35840