Learn More
Medchemexpress LLC 3,4,5-Trimethoxy-N-(2-methoxyethyl)-N-(4-phenyl-2-thiazolyl)-benzamide | 461000-66-8 | MFCD03115431 | 98.9% | 428.50 | C22H24N2O5S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10336810MG
Encompass_Preferred
Eact is a small-molecule activator of the calcium-activated chloride channel TMEM16A (ANO1) that also directly activates TRPV1 channels in sensory nociceptors, producing itch, acute nociception, and thermal hypersensitivity. It is supplied as a purified white to off-white solid for research use with characterized physicochemical and solubility properties.
  • Selective activator of TMEM16A and TRPV1 channels.
  • High purity solid (≈98.9%) suitable for in vitro and in vivo studies.
  • Soluble in DMSO (50 mg/mL) and formulatable for in vivo use (≥2.5 mg/mL in 10% DMSO/90% corn oil).
  • Useful for electrophysiology and behavioral assays probing ion channel function.
  • Stable under recommended storage conditions for long-term use.
Catalog No. 50-002-44579