Learn More
Cayman Chemical 3 4 5Trismethoxybenzophenone
SDP
Supplier:  Cayman Chemical 1000418550 MG
Encompass_Preferred
An analog of trimethoxy resveratrol; inhibits proliferation of a variety of human tumor cell lines at 0.4-2 UG/ml
Catalog No. 50-289-1389