Learn More
Medchemexpress LLC 3-[4-(6-(3-(dimethylamino)propoxy)benzo[d]oxazol-2-yl)phenoxy]-N,N-dimethylpropan-1-amine | 1219962-49-8 | 98.1% | 397.51 g·mol-1 | C23H31N3O3 | 100MG
SDP
Supplier:  Medchemexpress LLC HY124603100MG
Encompass_Preferred
AT791 is a small-molecule inhibitor of Toll-like receptors 7 and 9 (TLR7 and TLR9) described in peer-reviewed literature. It is orally bioavailable, blocks TLR7/9-mediated signaling, and inhibits DNA-TLR9 interaction in vitro, making it useful for preclinical studies of innate immunity and autoimmune disease mechanisms.
  • Small-molecule inhibitor of TLR7 and TLR9.
  • Orally bioavailable, suitable for in vivo studies.
  • Inhibits DNA-TLR9 interaction in vitro.
  • Useful for preclinical research on innate immunity and autoimmunity.
  • Molecular formula C23H31N3O3; molecular weight 397.51 g·mol-1.
  • Typical storage: -20 °C for the powder.
Catalog No. 50-002-37160