Learn More
Medchemexpress LLC 3,4,6-tri-O-benzyl-D-galactal | 80040-79-5 | MFCD09039276 | 98.9% | 416.51 g/mol | C27H28O4 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW090103
Encompass_Preferred
3,4,6-Tri-O-benzyl-D-galactal is a protected sugar derivative used as an intermediate and reagent in glycobiology and carbohydrate chemistry. It is typically used for synthesis and protection-group strategies in oligosaccharide assembly and related research applications.
  • Protected sugar derivative suitable for synthetic chemistry applications.
  • High purity for reliable reaction outcomes.
  • Available in multiple pack sizes to support small-scale and preparative work.
  • Low moisture and stable under recommended storage conditions.
  • Well-characterized molecular weight and formula for accurate stoichiometry.
Catalog No. 50-002-08857