Learn More
Medchemexpress LLC 3,4,6-tri-O-benzyl-D-galactal | 80040-79-5 | MFCD09039276 | 98.9% | 416.51 g·mol⁻¹ | C27H28O4 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW090103500MG
Encompass_Preferred
3,4,6-Tri-O-benzyl-D-galactal is a protected galactal derivative used as a synthetic building block in carbohydrate chemistry and glycobiology research. It is applied in oligosaccharide synthesis and method development for selective glycosylation reactions.
  • Used as a building block in carbohydrate and glycan synthesis.
  • Provides benzyl protection for selective transformations.
  • Reported high purity supports analytical and synthetic use.
  • Available in laboratory-scale pack sizes, including 500 MG.
Catalog No. 50-002-08759