Learn More
Medchemexpress LLC 3,4,6-Tri-O-benzyl-D-glucal | 55628-54-1 | MFCD00061640 | 97.0% | 416.51 g/mol | C27H28O4 | 1 ML

Supplier: Medchemexpress LLC HYW06161610MM/1ML
Tri-O-benzyl-D-glucal is a benzyl-protected glucal used as a glycosyl donor and intermediate in carbohydrate synthesis and glycobiology research. It is supplied as a solid and as a ready-to-use 10 mM solution in DMSO (1 mL). The material has a reported catalog purity of 97% and a batch HPLC purity of 99.82% for batch 731460.
- chemical identity: 3,4,6-tri-O-benzyl-D-glucal; CAS 55628-54-1.
- molecular formula and weight: C27H28O4, 416.51 g/mol.
- forms available: solid, and 10 mM solution in DMSO (1 mL).
- typical use: glycosyl donor in carbohydrate synthesis and glycobiology research.
- purity: catalog-listed 97%; batch COA reports 99.82% (HPLC) for batch 731460.
- storage: powder -20 °C (up to 3 years); in solvent -80 °C (6 months) or -20 °C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.