Learn More
Medchemexpress LLC 3,4,7,8-Tetramethyl-1,10-phenanthroline | 1660-93-1 | 99.8% | 236.31 | 50 G
SDP
Supplier:  Medchemexpress LLC HY3451550G
Encompass_Preferred
3,4,7,8-Tetramethyl-1,10-phenanthroline (TMPhen) is an organic molecule commonly used as a ligand or catalyst. It has a wide range of applications in different fields, such as organometallic chemical reactions, electrochemical detection, and organic optoelectronic devices. Due to its excellent performance in fluorescent probes, biosensors and photocatalytic reactions, it has been widely used in research in the fields of chemistry and life sciences. It is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Functions as a ligand or catalyst
  • Used in organometallic chemical reactions
  • Applied in electrochemical detection
  • Valuable in organic optoelectronic devices
  • Excellent performance in fluorescent probes
  • Suitable for biosensors and photocatalytic reactions
  • Intended for research use only
Catalog No. 50-004-05016