Learn More
Medchemexpress LLC 3-(4-bromo-2-fluorophenyl)piperidine-2,6-dione | 2924092-66-8 | 98.6% | 286.10 g/mol | C11H9BrFNO2 | 250mg
SDP
Supplier:  Medchemexpress LLC HYW888970250MG
Encompass_Preferred
3-(4-Bromo-2-fluorophenyl)piperidine-2 6-dione is a CRBN-type E3 ubiquitin ligase ligand that can be used to prepare PROTAC
Catalog No. 50-002-68399