Learn More
Medchemexpress LLC 3-(4-bromo-7-fluoro-1-oxoisoindolin-2-yl)piperidine-2,6-dione | 2438239-34-8 | ≥97.0% | 341.13 g/mol | C13H10BrFN2O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW46022750MG
Encompass_Preferred
Lenalidomide-4-Br-7-fluoro is a synthetic analog of lenalidomide bearing bromine and fluorine substituents on the benzyl ring. Supplied as a solid research-use compound, it is used in chemical biology and medicinal chemistry to probe cereblon engagement and structure-activity relationships. The material is offered at high purity for analytical and biological studies.
  • Derivative of lenalidomide with bromo and fluoro substituents.
  • Used to study cereblon (CRBN) ligand interactions and degradation pathways.
  • Provided as a solid powder suitable for small-scale assays and synthesis.
  • High purity appropriate for analytical and biological experiments.
  • Available in small pack sizes for research screening workflows.
Catalog No. 50-002-84990