Learn More
Cayman Chemical 3 4-dehydro CIlostazol 25mg
SDP
Supplier:  Cayman Chemical 3507525 MG
Encompass_Preferred
An active metabolite of cilostazol; formed from cilostazol by CYP3A4 and CYP2C19
Catalog No. 50-291-1499