Learn More
Medchemexpress LLC 3,4-dibromo-Mal-PEG8-Boc | 2055198-02-0 | 98.0% | C27H45Br2NO12 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY141007100MG
Encompass_Preferred
3,4-Dibromo-Mal-PEG8-Boc is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are composed of two distinct ligands joined by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein. These molecules function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Composed of two distinct ligands joined by a linker
  • Targets an E3 ubiquitin ligase and a specific protein
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-55621