Learn More
Enzo Life Sciences 3,4 Dichloroisocoumarin (10mg). CAS: 51050-59-0
SDP
Supplier:  Enzo Life Sciences BMLPI1100010
Encompass_Preferred
A potent irreversible mechanism based serine protease inhibitor. Inhibits granzymes A, B, and H, cathepsin G, neutrophil elastase and proteinase 3. Inhibits activation of neutral sphingomyelinase and apoptosis triggered by daunorubicin as well as camptothecin-induced apoptosis in HL-60 cells. Inhibits migration of eosinophils through basement membrane components in vitro. Purity: ≥98% (Elemental Analysis). Long Term Storage: +4°C.
Catalog No. 50-201-1445