Learn More
Medchemexpress LLC 3',4'-Dihydroxyacetophenone | 1197-09-7 | 152.15 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN177510MM/1ML
Encompass_Preferred
3',4'-Dihydroxyacetophenone (3,4-DHAP), isolated from Picea Schrenkiana Needles, is recognized for its strong suppressive action against tyrosinase activity, demonstrating an IC50 of 10 μM. This compound is also characterized as a vasoactive agent and antioxidant, making it a subject of interest in various research applications.
  • Exhibits strong suppressive action against tyrosinase activity
  • Functions as a vasoactive agent
  • Possesses antioxidant properties
  • Isolated from natural sources
Catalog No. 50-004-15420