Learn More
Medchemexpress LLC 3',4'-Dihydroxyacetophenone | 1197-09-7 | 152.15 | 10 G
SDP
Supplier:  Medchemexpress LLC HYN177510G
Encompass_Preferred
3',4'-Dihydroxyacetophenone (3,4-DHAP), isolated from Picea Schrenkiana Needles, exhibits a strong suppressive action against tyrosinase activity, with an IC50 of 10 μM. It is also identified as a vasoactive agent and an antioxidant. This product is suitable for various research areas including cancer studies.
  • Exhibits strong suppressive action against tyrosinase activity
  • Functions as a vasoactive agent
  • Acts as an antioxidant
Catalog No. 50-004-15779