Learn More
Medchemexpress LLC 3',4'-Dihydroxyacetophenone | 1197-09-7 | 152.15 | 100 G
SDP
Supplier:  Medchemexpress LLC HYN1775100G
Encompass_Preferred
3',4'-Dihydroxyacetophenone (3,4-DHAP) is isolated from Picea Schrenkiana Needles. It demonstrates a strong suppressive action against tyrosinase activity with an IC50 of 10 μM, and also functions as a vasoactive agent and an antioxidant.
  • Isolated from Picea Schrenkiana needles
  • Suppresses tyrosinase activity
  • Acts as a vasoactive agent
  • Functions as an antioxidant
Catalog No. 50-004-15870