Learn More
Medchemexpress LLC 3',4'-Dihydroxyacetophenone | 1197-09-7 | 152.15 | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHYN1775
Encompass_Preferred
3',4'-Dihydroxyacetophenone (3,4-DHAP) is a compound isolated from Picea Schrenkiana Needles. It exhibits a strong suppressive action against tyrosinase activity with an IC50 of 10 μM. This compound also functions as a vasoactive agent and an antioxidant.
  • Isolated from Picea Schrenkiana Needles
  • Strong suppressive action against tyrosinase activity
  • IC50 of 10 μM for tyrosinase
  • Functions as a vasoactive agent
  • Functions as an antioxidant
  • Targets tyrosinase
  • Pathway: Metabolic enzyme/protease
Catalog No. 50-003-89608