Learn More
Medchemexpress LLC 3',4'-Dihydroxyacetophenone (3,4-DHAP) | 1197-09-7 | 152.15 | 5 G
SDP
Supplier:  Medchemexpress LLC HYN17755G
Encompass_Preferred
3',4'-Dihydroxyacetophenone (3,4-DHAP) is a chemical compound isolated from Picea Schrenkiana needles. It exhibits a strong suppressive action against tyrosinase activity, with an IC50 of 10 μM. It also functions as a vasoactive agent and an antioxidant.
  • Isolated from Picea Schrenkiana needles.
  • Exhibits strong suppressive action against tyrosinase activity.
  • Has an IC50 of 10 μM against tyrosinase.
  • Acts as a vasoactive agent.
  • Functions as an antioxidant.
Catalog No. 50-004-15421