Learn More
Medchemexpress LLC 3,4-Dihydroxybenzeneacetic acid | 102-32-9 | 168.15 | 50 MG

Supplier: Medchemexpress LLC HYW00108050MG
3,4-Dihydroxybenzeneacetic acid is the main neuronal metabolite of dopamine, used for research purposes. Cerebrospinal fluid (CSF) 3,4-Dihydroxybenzeneacetic acid (DOPAC) is derived from intra-neuronal metabolism of cytoplasmic dopamine and acts as a sensitive and specific biomarker of central dopamine deficiency.
- Appearance: Solid
- Color: Off-white to light brown
- Molecular Formula: C8H8O4
- Target: Endogenous metabolite, drug metabolite
- Biomarker of central dopamine deficiency
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.