Learn More
Medchemexpress LLC 3,4-Dihydroxybenzeneacetic acid-d5 | 60696-39-1 | 173.18 | 100MG
SDP
Supplier:  Medchemexpress LLC HYW001080S100MG
Encompass_Preferred
3,4-Dihydroxybenzeneacetic acid-d5 is a deuterium labeled form of 3,4-Dihydroxybenzeneacetic acid. It is recognized as the main neuronal metabolite of dopamine.
  • Deuterium labeled form
  • Main neuronal metabolite of dopamine
  • Target: Endogenous metabolite
  • Pathway: Metabolic enzyme/protease
Catalog No. 50-004-46329