Learn More
Apexbio Technology LLC 3,4-Dihydroxybenzoic acid 99-50-3 1g
SDP
Supplier:  Apexbio Technology LLC N24661000
Encompass_Preferred
3 4-Dihydroxybenzoic acid (CAS 99-50-3) is a naturally occurring phenolic compound present in the leaves of Stenoloma chusanum (L ) Ching It exhibits antioxidative properties through the scavenging of reactive oxygen species thereby mitigating oxidative stress in biological systems This molecule has been investigated for its role in modulating cellular redox balance making it valuable for studies on oxidative damage neuroprotection and inflammation Its applications extend to biochemical assays evaluating antioxidant potential and cellular protection mechanisms The compound should be stored in a sealed container under cool dry conditions to maintain stability
Catalog No. 50-000-94434