Learn More
Medchemexpress LLC 3',4'-Dihydroxyflavone | 4143-64-0 | 98.62% | 1 ML

Supplier: Medchemexpress LLC HY12716710MM/1ML
3',4'-Dihydroxyflavone is a flavonoid that exhibits antiallergic activity with an IC50 value of 30 μM, making it suitable for research related to type I allergy. This compound is an off-white to light yellow solid.
- Antiallergic activity (IC50: 30 μM).
- Inhibits human telomerase in HEK293 cell extracts.
- Inhibits oxidative burst in PMA-stimulated human neutrophils.
- Shows antiinflammatory activity in mouse RAW264.7 cells.
- Molecular formula: C15H10O4.
- Molecular weight: 254.24.
- Provided as a 10 mM solution in DMSO, ready for reconstitution.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.