Learn More
Medchemexpress LLC 3',4'-Dihydroxyflavonol | 6068-78-6 | 99.3% | 270.24 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11180450MG
Encompass_Preferred
3',4'-Dihydroxyflavonol (DiOHF) is an effective antioxidant that reduces superoxide and improves nitric oxide (NO) function in diabetic rat mesenteric arteries. This compound is for research use only.
  • Reduces superoxide.
  • Improves nitric oxide (NO) function in diabetic rat mesenteric arteries.
  • Preserves NO activity in the presence of elevated reactive oxygen species (ROS).
  • Reduces Nox2-dependent superoxide production in diabetes.
  • Prevents eNOS uncoupling to improve endothelial function.
  • Reduces vascular contraction through calcium desensitization.
  • Exhibits neuroprotective activity with an EC50 of 0.5 μM in HT-22 cells.
Catalog No. 50-004-39391