Learn More
Medchemexpress LLC 3,4-dimethoxybenzyl alcohol | 93-03-8 | MFCD00004638 | 100.0% | 168.19 | C9H12O3 | 100 G
SDP
Supplier:  Medchemexpress LLC HY107858100G
Encompass_Preferred
Veratryl alcohol (3,4-dimethoxybenzyl alcohol) is an aromatic benzyl alcohol used as a research reagent and as a substrate for lignin peroxidase. It is a colorless to light-yellow liquid with molecular formula C9H12O3, molecular weight 168.19, and CAS 93-03-8. Commonly supplied for laboratory enzymatic and synthesis applications with documented analytical characterization.
  • Used as a substrate for lignin peroxidase and related enzymatic assays.
  • High purity suitable for analytical and research applications.
  • Stable aromatic alcohol for reagent and synthesis work.
  • Liquid form simplifies handling and solution preparation.
  • Characterized by documented analytical data (GC, LCMS).
Catalog No. 50-002-40822