Learn More
Medchemexpress LLC 3,4-dimethoxybenzyl alcohol | 93-03-8 | MFCD00004638 | 100.0% | 168.19 g/mol | C9H12O3 | 25 G

Supplier: Medchemexpress LLC HY10785825G
Veratryl alcohol (3,4-dimethoxybenzyl alcohol) is a research-grade small molecule used as a substrate in enzymatic assays and as a biochemical stabilizer. It is employed in studies of lignin degradation, enzyme activity, and microbial quorum sensing, and is supplied with high purity for laboratory research.
- Used as a substrate for lignin peroxidase assays.
- Acts as a stabilizer for manganese-dependent peroxidases.
- Reported quorum-sensing inhibitor and antibacterial activity.
- High purity (GC) - 99.98%.
- Molecular formula C9H12O3, molecular weight 168.19 g/mol.
- Recommended storage: pure form at -20°C (up to 3 years) or 4°C (up to 2 years).
- Available in 25 G laboratory pack.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.