Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3,4-dimethoxybenzyl alcohol | 93-03-8 | MFCD00004638 | 100.0% | 168.19 g·mol⁻¹ | C9H12O3 | 50 G
SDP

Supplier:  Medchemexpress LLC HY10785850G

Encompass_Preferred

Veratryl alcohol (3,4-dimethoxybenzyl alcohol) is an aromatic benzyl alcohol reagent used as a substrate and stabilizer in studies of lignin peroxidase and other oxidative enzymes. It is supplied as a high-purity solid suitable for enzymology assays, metabolic studies, and organic synthesis.

  • High purity for analytical and preparative work.
  • Molecular formula C9H12O3 and molecular weight 168.19 g·mol⁻¹.
  • Solid reagent suitable for bench-scale handling and storage.
  • Useful substrate for lignin peroxidase and other enzyme assays.
  • Stable under recommended storage conditions when refrigerated or frozen.
  • Soluble in common organic solvents for reaction and assay preparation.

Catalog No. 50-002-42770


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.