Learn More
Medchemexpress LLC 3,4-dimethoxybenzyl alcohol | 93-03-8 | MFCD00004638 | ≥98.0% | 168.19 g/mol | C9H12O3 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1078581G
Encompass_Preferred
Veratryl alcohol (3,4-dimethoxybenzyl alcohol) is an aromatic small-molecule research reagent used in enzymatic and biochemical studies. It functions as a substrate for lignin-degrading peroxidases and can stabilize oxidative reactions, making it useful in assay development and enzymology research. The compound is provided as a high-purity reagent for laboratory use.
  • Used as a substrate for lignin peroxidase assays.
  • Acts as a stabilizer in hydrogen peroxide-containing reactions.
  • High purity suitable for analytical and preparative applications.
  • Molecular formula C9H12O3 and molecular weight 168.19 g/mol.
  • Suitable for biochemical research and enzymology studies.
Catalog No. 50-002-40283