Learn More
AdipoGen 3,4-Dimethoxychalcone (10 mg)
SDP
Supplier:  AdipoGen AGCN20531M010
Encompass_Preferred
Chemical. CAS: 5416-71-7. Formula: C17H16O3. MW: 268.3. Synthetic. Caloric restriction mimetic (CRM). Caloric restriction mimetics (CRMs) are natural or synthetic compounds that mimic the health-promoting and longevity-extending effects of caloric restriction. CRMs provoke the deacetylation of cellular proteins coupled to an increase in autophagic flux in the absence of toxicity. Induces the deacetylation of cytoplasmic proteins and stimulates autophagic flux, requiring transcription factor EB (TFEB)- and E3 (TFE3)-dependent gene transcription and mRNA translation to trigger autophagy. Stimulates the translocation of TFEB and TFE3 into nuclei both in vitro and in vivo, in hepatocytes and cardiomyocytes. Consequently induces autophagy in vitro and in vivo, mediates autophagy-dependent cardioprotective effects and improves the efficacy of anticancer chemotherapy in vivo.
Catalog No. 50-205-3709