Learn More
Medchemexpress LLC 3,4-Dimethoxyphenol | 2033-89-8 | 154.16 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN178010MM/1ML
Encompass_Preferred
3,4-Dimethoxyphenol is a plant-derived phenylpropanoid compound that can be used as a whitening agent in cosmetics. It exhibits broad-spectrum antimicrobial activity and has tyrosinase-inhibiting activity. It also possesses a potent antioxidant effect and can be used for the study of infection.
  • Can be used as a whitening agent in cosmetics
  • Exhibits broad-spectrum antimicrobial activity
  • Has tyrosinase-inhibiting activity
  • Possesses a potent antioxidant effect
  • Can be utilized for the study of infection
Catalog No. 50-004-15550