Learn More
Medchemexpress LLC 3,4-Dimethoxyphenol | 2033-89-8 | 154.16 | 100 G
Supplier: Medchemexpress LLC HYN1780100G
3,4-Dimethoxyphenol is a plant-derived phenylpropanoid compound used as a whitening agent in cosmetics. It exhibits broad-spectrum antimicrobial activity, has tyrosinase-inhibiting activity, and shows a potent antioxidant effect.
- Plant-derived phenylpropanoid compound
- Used as a whitening agent in cosmetics
- Exhibits broad-spectrum antimicrobial activity
- Has tyrosinase-inhibiting activity
- Shows a potent antioxidant effect
- Can be used for the study of infection