Learn More
Medchemexpress LLC 3,4-Dimethoxyphenol | 2033-89-8 | 154.16 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYN1780
Encompass_Preferred
3,4-Dimethoxyphenol is a plant-derived phenylpropanoid compound with multiple applications. It acts as a whitening agent in cosmetics and demonstrates broad-spectrum antimicrobial activity. Additionally, it exhibits tyrosinase-inhibiting activity and possesses a potent antioxidant effect, having been isolated from bacterial fermentation broth. It is also used for studying infections.
  • Plant-derived phenylpropanoid compound.
  • Can be used as a whitening agent in cosmetics.
  • Exhibits broad-spectrum antimicrobial activity.
  • Has tyrosinase-inhibiting activity.
  • Potent antioxidant effect isolated from bacterial fermentation broth.
  • Can be used for the study of infection.
Catalog No. 50-003-89426