Learn More
Medchemexpress LLC 3,4-Dimethoxyphenol | 2033-89-8 | 154.16 | 5 G
SDP
Supplier:  Medchemexpress LLC HYN17805G
Encompass_Preferred
3,4-Dimethoxyphenol is a plant-derived phenylpropanoid compound that can be used as a whitening agent in cosmetics. It exhibits broad-spectrum antimicrobial activity, tyrosinase-inhibiting activity, and potent antioxidant effects. It can be used for the study of infection.
  • Plant-derived phenylpropanoid compound.
  • Can be used as a whitening agent in cosmetics.
  • Exhibits broad-spectrum antimicrobial activity.
  • Has tyrosinase-inhibiting activity.
  • Potent antioxidant effect isolated from bacterial fermentation broth.
  • Can be used for the study of infection.
  • Appearance: Solid.
  • Color: Off-white to yellow.
  • Initial source: Microorganisms (Aspergillus fumigatus).
Catalog No. 50-004-16323