Learn More
Cayman Chemical 3 4-DmAhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1112810 MG
Encompass_Preferred
An MDMA analog that appears to act as a serotonin-norepinephrine-dopamine releasing agent binding to and inhibiting uptake at noradrenalin and serotonin transporters (Kis = 22.8 and 7.7 μM and IC50s = 253.4 and 108 μM, respectively); significantly less potent than MDMA; intended for forensic and research applications
Catalog No. 50-295-1236