Learn More
MSC Proto J5649BL 3/4" Drive Pear Head Standard Ratchet Black Oxide Finish, 20" Overall Length, 24 Gear Teeth, Standard Knurled Handle, Standard Head
Supplier:  MSC 84968510
Encompass
Proto J5649BL 3/4" Drive Pear Head Standard Ratchet Black Oxide Finish, 20" Overall Length, 24 Gear Teeth, Standard Knurled Handle, Standard Head
Catalog No. 50-129-7908