Learn More
Medchemexpress LLC 3-(4-Fluoro-1-oxoisoindolin-2-yl)piperidine-2,6-dione | 2359705-88-5 | MFCD31556181 | 99.9% | 262.24 g/mol | C13H11FN2O3 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0392331G
Encompass_Preferred
Lenalidomide-F is an E3 ligase ligand that recruits cereblon and is used as a building block in PROTAC development and target protein degradation research. It is supplied as a research-grade reagent for chemical biology, degradation studies, and analytical applications.
  • E3 ligase ligand that recruits cereblon.
  • Used in PROTAC design to induce targeted protein degradation.
  • Reported activity in FLT3 degrader applications (DC50 = 0.64 nM).
  • High purity (99.9%) suitable for analytical and synthesis work.
  • Molecular formula C13H11FN2O3, molar mass 262.24 g/mol.
Catalog No. 50-002-99811