Learn More
Medchemexpress LLC 3-(4-fluorophenyl)propanoic acid | 459-31-4 | MFCD00060327 | 99.9% | 168.16 g·mol⁻¹ | C9H9FO2 | 100 G

Supplier: Medchemexpress LLC HYW028185100G
3-(4-Fluorophenyl)propanoic acid is an aromatic propanoic acid used as a biochemical reagent and synthetic intermediate in medicinal chemistry and life-science research. It functions as a fluorinated phenylpropionic acid building block for organic synthesis and compound modification, and is supplied with supporting analytical documentation for research use.
- High purity: 99.9% (reported 99.88%).
- Molecular weight 168.16 g·mol⁻¹.
- Chemical formula C9H9FO2.
- Appearance: white to off-white solid.
- Recommended storage: powder at -20°C (3 years) or 4°C (2 years).
- Certificate of analysis and safety data sheet available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.