Learn More
Medchemexpress LLC 3-(4-fluorophenyl)propanoic acid | 459-31-4 | MFCD00060327 | 99.9% | 168.16 g/mol | C9H9FO2 | 250 G

Supplier: Medchemexpress LLC 250GHYW028185
3-(4-Fluorophenyl)propanoic acid (CAS 459-31-4) is an organic biochemical reagent supplied as a white to off-white solid. It is used as a synthetic building block and intermediate in life-science and organic chemistry research. The product is provided with lot-specific documentation and intended for laboratory research use only.
- High purity (99.9%) suitable for research applications.
- White to off-white solid, easy to handle.
- Molecular formula C9H9FO2; molecular weight 168.16 g/mol.
- Available in multiple pack sizes, including 250 g.
- COA and SDS available for lot-specific documentation.
- Intended for research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.