Learn More
Cayman Chemical 3 4-MDPAhydrochlorIde 50mg
SDP
Supplier:  Cayman Chemical 1397250 MG
Encompass_Preferred
A lesser known psychoactive substituted amphetamine of the phenethylamine class that has been used as an internal standard for qualitative and quantitative analyses of hallucinogenic designer drugs in biological samples; intended for forensic and research purposes
Catalog No. 50-295-4594