Learn More
Medchemexpress LLC 3-[4-methyl-2-[(Z)-(2-oxo-1H-indol-3-ylidene)methyl]-1H-pyrrol-3-yl]propanoic acid | 215543-92-3 | MFCD08235144 | >97.0% | 296.32 Da | C17H16N2O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1040710MG
Encompass_Preferred
SU 5402 is a small-molecule, multi-targeted receptor tyrosine kinase inhibitor used in research to block VEGFR2, FGFR1, and PDGFRβ signaling. It exhibits nanomolar potency against VEGFR2 and FGFR1 and sub-micromolar activity against PDGFRβ, and is commonly used to study angiogenesis and growth factor pathways in vitro.
  • Potent inhibition of VEGFR2, FGFR1, and PDGFRβ (IC50: 20 nM, 30 nM, 510 nM).
  • Useful for studying angiogenesis and growth factor signaling in cell-based assays.
  • Cell-permeable, ATP-competitive tyrosine kinase inhibitor.
  • Molecular weight 296.32 Da and formula C17H16N2O3.
  • Provided as a high-purity research reagent suitable for biochemical and cellular studies.
Catalog No. 50-002-42563