Learn More
Medchemexpress LLC 3,4-Methylenedioxycinnamic acid | 2373-80-0 | 99.9% | 192.17 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00999310MM/1ML
Encompass_Preferred
3,4-Methylenedioxycinnamic acid is an inhibitor of the 4-hydroxycinnamoyl-CoA ligase (4CL) with a Ki of 100 μM. It exhibits a Km value for the substrate ferulic acid of 4CL of 8.4 μM. This compound increases the formation of soluble phenolics, particularly vanillic acid, and demonstrates larvicidal activity.
  • Inhibits 4-hydroxycinnamoyl-CoA ligase (4CL).
  • Increases the formation of soluble phenolics.
  • Demonstrates larvicidal activity.
  • For research use only.
Catalog No. 50-004-53636