Learn More
Medchemexpress LLC 3,4-Methylenedioxycinnamic acid | 2373-80-0 | MFCD00005837 | 99.7% | 192.17 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0099935G
Encompass_Preferred
3,4-Methylenedioxycinnamic acid is an inhibitor of 4-hydroxycinnamoyl-CoA ligase (4CL) with a Ki of 100 μM. It exhibits a Km value for the substrate ferulic acid of 4CL at 8.4 μM. This compound increases the formation of soluble phenolics, particularly vanillic acid, and demonstrates larvicidal activity.
  • Inhibitor of 4-hydroxycinnamoyl-CoA ligase (4CL).
  • Increases the formation of soluble phenolics, particularly vanillic acid.
  • Demonstrates larvicidal activity against Aedes aegypti.
  • Exhibits cellular effects across multiple cell lines (HeLa, HepG2, MDCK, MT4, Oocyte, RAW264.7).
  • Shows in vivo activity with mild central nervous system effects at 2000 mg/kg in female Swiss mice.
  • Histological analysis showed no structural signs of toxicity in kidneys, liver, spleen, and lungs in tested animal models.
Catalog No. 50-004-52507