Learn More
Medchemexpress LLC 3-(4-Pyridyl)indole (Rockout) | 7272-84-6 | 99.9% | 194.23 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11236210MG
Encompass_Preferred
3-(4-Pyridyl)indole (Rockout) is a Rho-kinase (ROCK) inhibitor.
  • Inhibits Rho-kinase (ROCK).
  • Potency with an IC50 of 25 μM.
  • Inhibits blebbing.
  • Causes dissolution of actin stress fibers in a wound healing assay.
Catalog No. 50-004-38638