Learn More
Chemglass Life Sciences 3/4 Sanitary Genderless AseptiQuik S Connector, High Temperature
SDP
Supplier:  Chemglass Life Sciences CLS91311012HT
Encompass
Material: Polycarbonate, USP Class VI, ADCF. Material Finish: Molded White. Pressure Range: Up to 60 psi, 4. 1 bar. Color: White with white tab covers. Mounting Option: Free Floating. Supplied Non-Sterile: Autoclave and Gamma Radiation. Temperature Range: 39 - 104 ° F (4 - 40 ° C). Seal Material: Silcone, Platinum-cured, USP Class VI, ADCF. Valve Option: Non-Valved.
Catalog No. 50-009-80703