Learn More
Alkali Scientific T150 U-Shaped Cell Culture Flask, Plug Seal, Non-Treated
SDP
Supplier:  Alkali Scientific T150UNT
Encompass
A brand new specification, T150 U-shaped Canted Neck Cell Culture Flask, is added to the collection, which deviates from the regular design. Without influencing the regular usage, the bottleneck is enlarged, allowing the Pipettes or cell scrapers to reach every corner of the flask freely and flexibly. The unique U-shaped body is easier to hold and enhances the utilization rate of the bottom. A brand new specification, T150 U-shaped Canted Neck Cell Culture Flask, is added to the collection, which deviates from the regular design. Without influencing the regular usage, the bottleneck is enlarged, allowing the Pipettes or cell scrapers to reach every corner of the flask freely and flexibly. The unique U-shaped body is easier to hold and enhances the utilization rate of the bottom.
Catalog No. 50-302-9242