Learn More
Cayman Chemical 3 4DIhydroquInolIn2 1Hon 10g
SDP
Supplier:  Cayman Chemical 3170110 G
Encompass_Preferred
A building block; has been used in the synthesis of compounds with antidepressant and anticonvulsant activities, as well as dual ligands of σ receptors and GluN2b that have antioxidant and neuroprotective properties
Catalog No. 50-299-2605