Learn More
Accuflow Systems 3/4IN TC CAP PP CLASS VI 10/PK
Supplier:  Accuflow Systems PEC050075FC
Encompass_Preferred
1/2"-3/4" Polypropylene Bio End Caps are designed for use in GMP Manufacturing and laboratories. These caps can be autoclavable as well as gama irradiated. Constructed of USP Class VI Polypropylene. Designed for Sanitary Tri clamp applications.Max working pressure, 150psi @ 70� F, Pack of 10
Catalog No. 50-110-8238